2,5-Dichlorphenol ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 2,5-DCP

B24787.14 B24787.22 B24787.36
B24787.14EA 24.6 EUR
B24787.14 B24787.22 B24787.36
2,5-Dichlorphenol ≥98%
2,5-Dichlorphenol

Purity: 98% CAS No.: 583-78-8 Molecular Formula: C6H4Cl2O Formula Weight: 163.00 Odour: Phenol-like Melting Point: 55-58° Boiling Point: 211° Beilstein Reference: 1907692 UN No.: 2020 MDL No.: MFCD00002174

Formel: C₆H₄Cl₂O
Siedepunkt: 211 °C
Schmelzpunkt: 55…58 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00002174
CAS-Nummer: 583-78-8
EINECS: 209-520-4
UN: 2020
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR