2-Chlor-α-methylbenzylalkohol 96%

Lieferant: Thermo Fisher Scientific

Synonyme: 1-(2-Chlorophenyl)ethanol, o-Chloro-α-methylbenzyl alcohol, 2-Chlorophenyl methyl carbinol

B24800.06 B24800.14 AB146737.0005 AB146737.0025
B24800.06EA 35.4 EUR
B24800.06 B24800.14 ABCRAB146737.0005 ABCRAB146737.0025
2-Chlor-α-methylbenzylalkohol 96%
2-Chlor-α-methylbenzylalkohol

Purity: 96% CAS No.: 13524-04-4 Molecular Formula: C8H9ClO Formula Weight: 156.61 Refractive Index: 1.5450 Boiling Point: 121-123°/14mm MDL No.: MFCD00041037

Formel: C₈H₉ClO
Molecular Weight: 156.61 g/mol
Siedepunkt: 121…123 °C (14 mmHg)
Lagertemperatur: Raumtemperatur
MDL: MFCD00041037
CAS-Nummer: 13524-04-4
EINECS: 236-868-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR