6-Hydroxyhexanoic acid 95% may cont. variable amounts of dimer

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 6-Hydroxycaproic acid

B24857.03 B24857.06 B24857.14
B24857.03EA 54.1 EUR
B24857.03 B24857.06 B24857.14
6-Hydroxyhexanoic acid 95% may cont. variable amounts of dimer
6-Hydroxyhexanoic acid

Purity: 95% CAS No.: 1191-25-9 Molecular Formula: C6H12O3 Formula Weight: 132.16 Storage Temperature: -30°C to -10°C Flash Point: 110°(230°F) Refractive Index: 1.4560 MDL No.: MFCD00046560

Formel: C₆H₁₂O₃
Molecular Weight: 132.159 g/mol
Dichte: 1.08
Flammpunkt: >110 °C (230 °F)
MDL: MFCD00046560
CAS-Nummer: 1191-25-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR