1,2:3,4-Di-O-isopropylidene-ɑ-D-galactopyranose 95%

Lieferant: Thermo Fisher Scientific

B24899.06 B24899.14
B24899.06EA 40.2 EUR
B24899.06 B24899.14
1,2:3,4-Di-O-isopropylidene-ɑ-D-galactopyranose 95%
1,2:3,4-Di-O-isopropylidene-alpha-D-galactopyranose

Syn: Diacetone-D-galactose Purity: 95% CAS No.: 4064-06-6 Molecular Formula: C12H20O6 Formula Weight: 260.29 MDL No.: MFCD00063225

Formel: C₁₂H₂₀O₆
Dichte: 1,143
Lagertemperatur: Raumtemperatur
MDL: MFCD00063225
CAS-Nummer: 4064-06-6
EINECS: 223-771-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR