2-(4-Hydroxyphenyl)propionsäure ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 2-(4-Hydroxyphenyl)propionic Acid

B24938.09 B24938.18
B24938.09EA 59 EUR
B24938.09 B24938.18
2-(4-Hydroxyphenyl)propionsäure ≥98%
2-(4-Hydroxyphenyl)propionsäure

Purity: 98% CAS No.: 938-96-5 Molecular Formula: C9H10O3 Formula Weight: 166.18 Melting Point: 130-132° MDL No.: MFCD00128847

Formel: C₉H₁₀O₃
Schmelzpunkt: 130…132 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00128847
CAS-Nummer: 938-96-5
EINECS: 213-352-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR