5-(4-Morpholinyl)-2-nitrophenol ≥97%

Lieferant: Thermo Fisher Scientific
Achtung

B24950.03 B24950.06
B24950.03EA 30.6 EUR
B24950.03 B24950.06
5-(4-Morpholinyl)-2-nitrophenol ≥97%
5-(4-Morpholinyl)-2-nitrophenol
Formel: C₁₀H₁₂N₂O₄
Molecular Weight: 224.22 g/mol
Schmelzpunkt: 143…145 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00053057
CAS-Nummer: 175135-19-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR