5-(4-Morpholinyl)-2-nitrophenol ≥97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
B24950.03
B24950.06
B24950.03EA
30.6
EUR
B24950.03
B24950.06
5-(4-Morpholinyl)-2-nitrophenol ≥97%
5-(4-Morpholinyl)-2-nitrophenol
Formel:
C₁₀H₁₂N₂O₄ Schmelzpunkt: 143…145 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00053057 CAS-Nummer: 175135-19-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...