cis-5-Norbornene endo-2,3-dicarboxylic anhydride 97%

Lieferant: Thermo Fisher Scientific
Gefahr

B24951.06 B24951.14 B24951.22
B24951.06EA 32 EUR
B24951.06 B24951.14 B24951.22
cis-5-Norbornene endo-2,3-dicarboxylic anhydride 97%
cis-5-Norbornene endo-2,3-dicarboxylic anhydride

Purity: 97% CAS No.: 129-64-6 Molecular Formula: C9H8O3 Formula Weight: 164.16 Melting Point: 164-167° Beilstein Reference: 83657 MDL No.: MFCD00151106

Formel: C₉H₈O₃
Schmelzpunkt: 164…167 °C
Dichte: 1,417
Lagertemperatur: Raumtemperatur
MDL: MFCD00151106
CAS-Nummer: 129-64-6
EINECS: 204-957-7
Merck Index: 14,1796

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR