1,1-Dimethoxyaceton ≥97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Purity: 97+%
CAS No.: 6342-56-9
Molecular Formula: CH3COCH(OCH3)2
Formula Weight: 118.13
Flash Point: 34°(93°F)
Refractive Index: 1.3980
Melting Point: -57°
Boiling Point: 133-135°
Beilstein Reference: 1560557
UN No.: 1993
MDL No.: MFCD00008758
Formel:
C₅H₁₀O₃ Siedepunkt: 133…135 °C Schmelzpunkt: –57 °C Dichte: 0,998 Flammpunkt: 34 °C (93 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00008758 CAS-Nummer: 6342-56-9 EINECS: 228-735-4 UN: 1993 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...