Natriumphenoxid ≥98%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
Sodium phenylate
B25002.18
B25002.30
B25002.18EA
78.4
EUR
B25002.18
B25002.30
Natriumphenoxid ≥98%
Sodium phenoxide
Purity: 98%
CAS No.: 139-02-6
Molecular Formula: C6H5NaO
Formula Weight: 116.09
Odour: Phenol-like
Melting Point:
Formel:
C₆H₅NaO Schmelzpunkt: >300 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00013134 CAS-Nummer: 139-02-6 EINECS: 205-347-3 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...