N-Boc-L-phenylalaninol ≥99%

Lieferant: Thermo Fisher Scientific

B25019.03 B25019.06 B25019.14
B25019.03EA 23.4 EUR
B25019.03 B25019.06 B25019.14
N-Boc-L-phenylalaninol ≥99%
N-Boc-L-phenylalaninol

Purity: 99% CAS No.: 66605-57-0 Molecular Formula: C14H21NO3 Formula Weight: 251.33 Storage Temperature: 2°C to 8°C Melting Point: 95-98° Beilstein Reference: 3652004 Optical Rotation: -27.5° ± 2.5° (c=1 in methanol) MDL No.: MFCD00076976

Formel: C₁₄H₂₁NO₃
Schmelzpunkt: 95…98 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00076976
CAS-Nummer: 66605-57-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR