Formaldoxime trimer hydrochloride 95%

Lieferant: Thermo Fisher Scientific
Gefahr

B25063.09
B25063.09EA 106 EUR
B25063.09
Formaldoxime trimer hydrochloride 95%
Formaldoxime trimer hydrochloride

Purity: 95% CAS No.: 6286-29-9 Molecular Formula: C3H9N3O3.HCl Formula Weight: 171.57 Melting Point: ca 128° dec. Beilstein Reference: 3690722 UN No.: UN2811 MDL No.: MFCD00012778

Formel: C₃H₉N₃O₃·HCl
Schmelzpunkt: ∼ 128 °C (Zersetzung)
Lagertemperatur: Raumtemperatur
MDL: MFCD00012778
CAS-Nummer: 6286-29-9
EINECS: 222-440-4
ADR: 6.1,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR