trans-4-Aminocyclohexanol Hydrochlorid 97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
B25070.06
B25070.14
B25070.22
B25070.06EA
27.1
EUR
B25070.06
B25070.14
B25070.22
trans-4-Aminocyclohexanol Hydrochlorid 97%
trans-4-Aminocyclohexanol Hydrochlorid
Purity: 97%
CAS No.: 50910-54-8
Molecular Formula: C6H13NO.HCl
Formula Weight: 151.64
Melting Point: 225-230°
Beilstein Reference: 3909294
Formel:
C₆H₁₄ClNO Schmelzpunkt: 225…230 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00012566 CAS-Nummer: 50910-54-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...