4,4'-Dimethylbiphenyl ≥99%

Lieferant: Thermo Fisher Scientific

Synonyme: 4,4'-Bitolyl, 1-Methyl-4-(4-methylphenyl)benzol

B25089.03 B25089.06 B25089.14
B25089.03EA 40.5 EUR
B25089.03 B25089.06 B25089.14
4,4'-Dimethylbiphenyl ≥99%
4,4'-Dimethylbiphenyl

Purity: 99% CAS No.: 613-33-2 Molecular Formula: C14H14 Formula Weight: 182.27 Melting Point: 118-122° Boiling Point: 295° MDL No.: MFCD00008545

Formel: C₁₄H₁₄
Molecular Weight: 182.27 g/mol
Siedepunkt: 295 °C
Schmelzpunkt: 118…122 °C
Dichte: 1.102 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00008545
CAS-Nummer: 613-33-2
EINECS: 210-337-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR