3-Methyl-3-oxetanmethanol ≥97%
Lieferant: Thermo Fisher Scientific
B25111.14
B25111.22
B25111.14EA
62.7
EUR
B25111.14
B25111.22
3-Methyl-3-oxetanmethanol ≥97%
3-Methyl-3-oxetanmethanol
Purity: 97%
CAS No.: 3143-02-0
Molecular Formula: C5H10O2
Formula Weight: 102.13
Storage Temperature: 2°C to 8°C
Flash Point: 98°(208°F)
Refractive Index: 1.4460
Boiling Point: 79-80°/40mm
Beilstein Reference: 102773
MDL No.: MFCD00010273
Formel:
C₅H₁₀O₂ Siedepunkt: 79…80 °C (40 mmHg) Dichte: 1,024 Flammpunkt: 98 °C (208 °F) |
MDL:
MFCD00010273 CAS-Nummer: 3143-02-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...