2,3-Pyridindicarbonsäureanhydrid ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

B25118.06 B25118.14 B25118.22
B25118.06EA 30.9 EUR
B25118.06 B25118.14 B25118.22
2,3-Pyridindicarbonsäureanhydrid ≥98%
2,3-Pyridindicarbonsäureanhydrid

Purity: 98% CAS No.: 699-98-9 Molecular Formula: C7H3NO3 Formula Weight: 149.11 Melting Point: 134-138° Beilstein Reference: 125111

Formel: C₇H₃NO₃
Siedepunkt: 360 °C (1013 hPa)
Schmelzpunkt: 134…138 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00005915
CAS-Nummer: 699-98-9
EINECS: 211-834-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR