(R)-(-)-1,3-Butandiol ≥98%

Lieferant: Thermo Fisher Scientific

B25124.03 B25124.06
B25124.03EA 59.6 EUR
B25124.03 B25124.06
(R)-(-)-1,3-Butandiol ≥98%
(R)-(-)-1,3-Butandiol

Purity: 98% CAS No.: 6290-03-5 Molecular Formula: C4H10O2 Formula Weight: 90.12 Flash Point: 121°(249°F) Refractive Index: 1.4400 Odour: Weak Boiling Point: 203-204° Beilstein Reference: 1718944 Optical Rotation: -30° (neat) MDL No.: MFCD00064277

Formel: C₄H₁₀O₂
Siedepunkt: 203…204 °C
Dichte: 1,005
Flammpunkt: 121 °C (249 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00064277
CAS-Nummer: 6290-03-5
EINECS: 228-532-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR