4,4'-Dibromoctafluorbiphenyl (DBOB) ≥99%

Lieferant: Thermo Scientific

Synonyme: 4,4-Dibromooctafluorobiphenyl

B25126.03
B25126.03EA 64.6 EUR
B25126.03
4,4'-Dibromoctafluorbiphenyl (DBOB) ≥99%
4,4'-Dibromoctafluorbiphenyl (DBOB)

Purity: 99% CAS No.: 10386-84-2 Molecular Formula: C12Br2F8 Formula Weight: 455.94 Melting Point: 113-115° UN No.: 3152 MDL No.: MFCD00000310

Formel: C₁₂Br₂F₈
Molecular Weight: 455,94 g/mol
Schmelzpunkt: 113…115 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00000310
CAS-Nummer: 10386-84-2
EINECS: 233-847-1
ADR: 9,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR