1,3-Di-o-tolylguanidin ≥99%

Lieferant: Thermo Fisher Scientific
Achtung

B25127.14 B25127.22 B25127.36
B25127.14EA 23.6 EUR
B25127.14 B25127.22 B25127.36
1,3-Di-o-tolylguanidin ≥99%
1,3-Di-o-tolylguanidin

Purity: 99% CAS No.: 97-39-2 Molecular Formula: C15H17N3 Formula Weight: 239.32 Melting Point: 176-178° UN No.: 2811 MDL No.: MFCD00008513

Formel: C₁₅H₁₇N₃
Schmelzpunkt: 176…178 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00008513
CAS-Nummer: 97-39-2
EINECS: 202-577-6
UN: 2811
ADR: ,

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR