N-(6-Bromhexyl)phthalimid ≥97%
Lieferant: Thermo Fisher Scientific
B25128.06
B25128.14
B25128.22
B25128.06EA
52.6
EUR
B25128.06
B25128.14
B25128.22
N-(6-Bromhexyl)phthalimid ≥97%
N-(6-Bromhexyl)phthalimid
Purity: 97%
CAS No.: 24566-79-8
Molecular Formula: C14H16BrNO2
Formula Weight: 310.19
Odour: Odourless
Melting Point: 54-60°
Boiling Point: 186-188°/0.4mm
Beilstein Reference: 186875
MDL No.: MFCD00023098
Formel:
C₁₄H₁₆BrNO₂ Siedepunkt: 186…188 °C (0,4 mmHg) Schmelzpunkt: 54…60 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00023098 CAS-Nummer: 24566-79-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...