4-(Chlormethyl)-1,2-diphenylethan ≥98%

Lieferant: Thermo Fisher Scientific
Gefahr

B25222.03 B25222.06 B25222.14
B25222.03EA 36.5 EUR
B25222.03 B25222.06 B25222.14
4-(Chlormethyl)-1,2-diphenylethan ≥98%
4-(Chlormethyl)-1,2-diphenylethan

Purity: 98% CAS No.: 80676-35-3 Molecular Formula: C15H15Cl Formula Weight: 230.73 Melting Point: 38-42° UN No.: 1759 MDL No.: MFCD02093918

Formel: C₁₅H₁₅Cl
Schmelzpunkt: 38…42 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD02093918
CAS-Nummer: 80676-35-3
ADR: 8,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR