5-Brom-2-hydroxypyridin ≥97%
Lieferant: Thermo Fisher Scientific
B25267.06
B25267.14
B25267.06EA
25
EUR
B25267.06
B25267.14
5-Brom-2-hydroxypyridin ≥97%
5-Brom-2-hydroxypyridin
Purity: 97%
CAS No.: 13466-38-1
Molecular Formula: C5H4BrNO
Formula Weight: 174.00
Melting Point: 178-182°
Beilstein Reference: 108751
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...