3-Nitrophthalonitril ≥99%

Lieferant: Thermo Fisher Scientific
Achtung

B25269.06 B25269.14
B25269.06EA 99.8 EUR
B25269.06 B25269.14
3-Nitrophthalonitril ≥99%
3-Nitrophthalonitril

Purity: 99% CAS No.: 51762-67-5 Molecular Formula: C8H3N3O2 Formula Weight: 173.13 Melting Point: 162-165° Beilstein Reference: 2263686

Formel: C₈H₃N₃O₂
Schmelzpunkt: 162…165 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00191558
CAS-Nummer: 51762-67-5
UN: 3276
ADR: 6.1,I

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR