Kupfer(II)hexafluoracetylacetonat Hydrat ≥98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
Kupfer(II)Hexafluoro-2,4-pentanedionat Hydrat
B25284.03
B25284.06
B25284.14
B25284.03EA
33.5
EUR
B25284.03
B25284.06
B25284.14
Kupfer(II)hexafluoracetylacetonat Hydrat ≥98%
Kupfer(II)hexafluoracetylacetonat Hydrat
Purity: 98%
CAS No.: 155640-85-0
Molecular Formula: C10H2CuF12O4.xH2O
Formula Weight: 477.64(anhy)
Melting Point: ca 100°
Boiling Point: 220°
MDL No.: MFCD00151019
Formel:
C₁₀H₂CuF₁₂O₄·H₂O Siedepunkt: 220 °C Schmelzpunkt: ∼ 100 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00151019 CAS-Nummer: 155640-85-0 EINECS: 238-850-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...