5-Methoxyindol ≥99%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
5-Methoxy-1H-indole, Indol-5-yl methyl ether
B25290.03
B25290.06
B25290.14
B25290.03EA
32.7
EUR
B25290.03
B25290.06
B25290.14
5-Methoxyindol ≥99%
5-Methoxyindol
Purity: 99%
CAS No.: 1006-94-6
Molecular Formula: C9H9NO
Formula Weight: 147.18
Storage Temperature: 2°C to 8°C
Melting Point: 52-55°
Boiling Point: 176-178°/17mm
Beilstein Reference: 116722
MDL No.: MFCD00005674
Formel:
C₉H₉NO Siedepunkt: 176…178 °C (17 mmHg) Schmelzpunkt: 52…55 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00005674 CAS-Nummer: 1006-94-6 EINECS: 213-745-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...