4-Bromomethyl-1,2-dihydroquinoline-2-one
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
B25305.06
B25305.06EA
53.2
EUR
B25305.06
4-Bromomethyl-1,2-dihydroquinoline-2-one
4-Bromomethyl-1,2-dihydroquinoline-2-one
CAS No.: 4876-10-2
Molecular Formula: C10H8BrNO
Formula Weight: 238.08
Melting Point: ca 254° dec.
UN No.: UN3261
MDL No.: MFCD03426152
Formel:
C₁₀H₈BrNO Schmelzpunkt: ∼ 254 °C (Zers.) Lagertemperatur: Raumtemperatur |
MDL:
MFCD03426152 CAS-Nummer: 4876-10-2 UN: 3261 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...