Mesitylenesulphonic acid dihydrate ≥98%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: 2,4,6-Trimethylbenzenesulphonic acid dihydrate

B25321.22
B25321.22EA 165 EUR
B25321.22
Mesitylenesulphonic acid dihydrate ≥98%
Mesitylenesulphonic acid dihydrate

Purity: 98+% CAS No.: 835617-36-2 Molecular Formula: C9H12O3S.2H2O Formula Weight: 236.29 (200.26anhy) Melting Point: 74-77° Beilstein Reference: 1961501 UN No.: UN2585 MDL No.: MFCD00149535

Formel: C₉H₁₂O₃S·2H₂O
Molecular Weight: 236.29 g/mol (200.26 anhy)
Schmelzpunkt: 74…77 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00149535
CAS-Nummer: 835617-36-2
EINECS: 222-379-3
ADR: 8 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR