Methyl-4-aminonicotinat 98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
Methyl 4-aminonicotinate
B25324.06
B25324.06EA
177
EUR
B25324.06
Methyl-4-aminonicotinat 98%
Methyl-4-aminonicotinat
Purity: 98%
CAS No.: 16135-36-7
Molecular Formula: C7H8N2O2
Formula Weight: 152.15
Melting Point: 172-174°
MDL No.: MFCD00955996
Formel:
C₇H₈N₂O₂ Molecular Weight: 152.15 g/mol Schmelzpunkt: 172…174 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00955996 CAS-Nummer: 16135-36-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...