4-Phenoxybenzonitril 96%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
B25364.04
B25364.09
B25364.04EA
26.2
EUR
B25364.04
B25364.09
4-Phenoxybenzonitril 96%
4-Phenoxybenzonitril
Purity: 96%
CAS No.: 3096-81-9
Molecular Formula: C13H9NO
Formula Weight: 195.22
Flash Point:
Formel:
C₁₃H₉NO Schmelzpunkt: 46…48 °C Flammpunkt: >110 °C (230 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00017346 CAS-Nummer: 3096-81-9 UN: 3439 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...