1-Chlor-2,6-dibrom-4-fluorbenzol ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 2-Chlor-1,3-dibrom-5-fluorbenzol

B25376.14
B25376.14EA 69.5 EUR
B25376.14
1-Chlor-2,6-dibrom-4-fluorbenzol ≥98%
1-Chloro-2,6-dibromo-4-fluorobenzene

Purity: 98% CAS No.: 179897-90-6 Molecular Formula: C6H2Br2ClF Formula Weight: 288.35 Melting Point: 84-87° MDL No.: MFCD00061122

Formel: C₆H₂Br₂ClF
Molecular Weight: 288.34 g/mol
Schmelzpunkt: 84…87 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00061122
CAS-Nummer: 179897-90-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR