Methyl-4-iod-3-methylbenzoat ≥98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
Methyl 4-iodo-3-methylbenzoate
B25395.22
B25395.14
B25395.06
B25395.22EA
502
EUR
B25395.22
B25395.14
B25395.06
Methyl-4-iod-3-methylbenzoat ≥98%
Methyl-4-iod-3-methylbenzoat
Purity: 98%
CAS No.: 5471-81-8
Molecular Formula: C9H9IO2
Formula Weight: 276.07
Melting Point: 59-62°
MDL No.: MFCD02683863
Formel:
C₉H₉IO₂ Molecular Weight: 276.07 g/mol Schmelzpunkt: 59…62 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD02683863 CAS-Nummer: 5471-81-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...