1-Chlor-3,5-dibrombenzol ≥98%

Lieferant: Thermo Scientific
Achtung

Synonyme: 3,5-Dibromochlorobenzene, 1,3-Dibromo-5-chlorobenzene

B25411.14
B25411.14EA 107 EUR
B25411.14
1-Chlor-3,5-dibrombenzol ≥98%
1-Chloro-3,5-dibromobenzene

Purity: 98% CAS No.: 14862-52-3 Molecular Formula: C6H3Br2Cl Formula Weight: 270.35 Melting Point: 95-98° MDL No.: MFCD00070765

Formel: C₆H₃Br₂Cl
Molecular Weight: 270,35 g/mol
Schmelzpunkt: 95…98 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00070765
CAS-Nummer: 14862-52-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR