Methyl-5-brompicolinat ≥98%

Lieferant: Thermo Scientific
Achtung

Synonyme: 5-Bromopyridine-2-carboxylic acid methyl ester, Methyl 5-bromopyridine-2-carboxylate, Methyl 5-bromopicolinate

B25415.06
B25415.06EA 153 EUR
B25415.06
Methyl-5-brompicolinat ≥98%
Methyl-5-brompicolinat

Purity: 98+% CAS No.: 29682-15-3 Molecular Formula: C7H6BrNO2 Formula Weight: 216.03 Melting Point: 99-103° MDL No.: MFCD04112493

Formel: C₇H₆BrNO₂
Molecular Weight: 216,03 g/mol
Schmelzpunkt: 99…103 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD04112493
CAS-Nummer: 29682-15-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR