3-Formylphenylboronsäure

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 3-Boronobenzaldehyd, Dihydroxy(3-formylphenyl)boran, (3-Formylphenyl)metaborsäure, 3-Formylbenzolboronsäure

B25437.03 B25437.06 B25437.14
B25437.03EA 40.3 EUR
B25437.03 B25437.06 B25437.14
3-Formylphenylboronsäure
3-Formylphenylboronsäure

CAS No.: 87199-16-4 Molecular Formula: C7H7BO3 Formula Weight: 149.94 Melting Point: 109-113° Beilstein Reference: 3030769 MDL No.: MFCD00161356

Formel: C₇H₇BO₃
Molecular Weight: 149.94 g/mol
Schmelzpunkt: 109…113 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00161356
CAS-Nummer: 87199-16-4

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR