Methyl-3-chlor-5-iodbenzoat ≥98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
Methyl 3-chloro-5-iodobenzoate
B25444.06
B25444.06EA
334
EUR
B25444.06
Methyl-3-chlor-5-iodbenzoat ≥98%
Methyl-3-chlor-5-iodbenzoat
Purity: 98%
CAS No.: 289039-85-6
Molecular Formula: C8H6ClIO2
Formula Weight: 296.49
Melting Point: 41-43°
MDL No.: MFCD00673018
Formel:
C₈H₆ClIO₂ Molecular Weight: 296,49 g/mol Schmelzpunkt: 41…43 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00673018 CAS-Nummer: 289039-85-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...