(±)-Indolin-2-carbonsäure 95%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Indoline-2-carboxylic acid

B25450.06 B25450.14
B25450.06EA 57.7 EUR
B25450.06 B25450.14
(±)-Indolin-2-carbonsäure 95%
(±)-Indolin-2-carbonsäure

Purity: 95% CAS No.: 78348-24-0 Molecular Formula: C9H9NO2 Formula Weight: 163.18 Melting Point: ca 168° dec. MDL No.: MFCD00010635

Formel: C₉H₉NO₂
Schmelzpunkt: ∼ 168 °C (Zers.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00010635
CAS-Nummer: 78348-24-0
EINECS: 278-899-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR