4-Chlor-2-iodanilin ≥98%

Lieferant: Thermo Fisher Scientific
Gefahr

B25457.14 B25457.06
B25457.14EA 352 EUR
B25457.14 B25457.06
4-Chlor-2-iodanilin ≥98%
4-Chlor-2-iodanilin

Purity: 98% CAS No.: 63069-48-7 Molecular Formula: C6H5ClIN Formula Weight: 253.47 Flash Point: 110(230°F) Melting Point: 41-44° UN No.: 2811 MDL No.: MFCD01863737

Formel: C₆H₅ClIN
Molecular Weight: 253.47 g/mol
Schmelzpunkt: 41…44 °C
Flammpunkt: >110 °C (230 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD01863737
CAS-Nummer: 63069-48-7
EINECS: 263-839-3
UN: 2811
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR