4-Brom-3-fluorbenzoesäure ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

B25475.06
B25475.06EA 39.2 EUR
B25475.06
4-Brom-3-fluorbenzoesäure ≥98%
4-Bromo-3-fluorobenzoic acid

Purity: 98+% CAS No.: 153556-42-4 Molecular Formula: C7H4BrFO2 Formula Weight: 219.01 Melting Point: 210-213° MDL No.: MFCD00672932

Formel: C₇H₄BrFO₂
Schmelzpunkt: 210…213 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00672932
CAS-Nummer: 153556-42-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR