4-Fluor-2-iodanilin ≥98%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
B25499.14
B25499.06
B25499.14EA
176
EUR
B25499.14
B25499.06
4-Fluor-2-iodanilin ≥98%
4-Fluor-2-iodanilin
Purity: 98%
CAS No.: 61272-76-2
Molecular Formula: C6H5FIN
Formula Weight: 237.02
UN No.: 2810
MDL No.: MFCD00042584
Formel:
C₆H₅FIN Molecular Weight: 237.02 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00042584 CAS-Nummer: 61272-76-2 UN: 2810 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...