4-Fluor-2-iodanilin ≥98%

Lieferant: Thermo Fisher Scientific
Gefahr

B25499.14 B25499.06
B25499.14EA 176 EUR
B25499.14 B25499.06
4-Fluor-2-iodanilin ≥98%
4-Fluor-2-iodanilin

Purity: 98% CAS No.: 61272-76-2 Molecular Formula: C6H5FIN Formula Weight: 237.02 UN No.: 2810 MDL No.: MFCD00042584

Formel: C₆H₅FIN
Molecular Weight: 237.02 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00042584
CAS-Nummer: 61272-76-2
UN: 2810
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR