5-Chlor-o-kresol ≥98%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: 4-Chlor-2-hydroxytoluol, 5-Chlor-2-methylphenol, 1-Chlor-3-hydroxy-4-methylbenzol

B25527.06 B25527.14
B25527.06EA 68.9 EUR
B25527.06 B25527.14
5-Chlor-o-kresol ≥98%
5-Chlor-o-kresol

Purity: 98% CAS No.: 5306-98-9 Molecular Formula: C7H7ClO Formula Weight: 142.59 Melting Point: 70-75° Boiling Point: 109-112°/15mm UN No.: 3437 MDL No.: MFCD00060672

Formel: C₇H₇ClO
Molecular Weight: 142.58 g/mol
Siedepunkt: 109…112 °C (15 mmHg)
Schmelzpunkt: 70…75 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00060672
CAS-Nummer: 5306-98-9
EINECS: 226-160-3
UN: 3437
ADR: 6.1 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR