1-Brom-2-chlor-4-fluorbenzol ≥98%

Lieferant: Thermo Scientific
Achtung

Synonyme: 4-Brom-3-chlor-1-fluorbenzol

B25538.06
B25538.06EA 38.9 EUR
B25538.06
1-Brom-2-chlor-4-fluorbenzol ≥98%
1-Bromo-2-chloro-4-fluorobenzene

Purity: 98% CAS No.: 110407-59-5 Molecular Formula: C6H3BrClF Formula Weight: 209.45 Refractive Index: 1.5520

Formel: C₆H₃BrClF
Molecular Weight: 209,44 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00142587
CAS-Nummer: 110407-59-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR