(R)-(-)-4-Penten-2-ol 95%

Lieferant: Thermo Fisher Scientific
Achtung

B25659.03 B25659.06
B25659.03EA 115 EUR
B25659.03 B25659.06
(R)-(-)-4-Penten-2-ol 95%
(R)-(-)-4-Penten-2-ol

Purity: 95% CAS No.: 64584-92-5 Molecular Formula: C5H10O Formula Weight: 86.13 Flash Point: 30°(86°F) Refractive Index: 1.4240 Boiling Point: 115-116° Optical Rotation: -5.2° (c=1 in chloroform) UN No.: 1987 MDL No.: MFCD03701536

Formel: C₅H₁₀O
Siedepunkt: 115…116 °C
Dichte: 0,837
Flammpunkt: 30 °C (86 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD03701536
CAS-Nummer: 64584-92-5
UN: 1987
ADR: 3,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR