(R)-(-)-4-Penten-2-ol 95%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
B25659.03
B25659.06
B25659.03EA
115
EUR
B25659.03
B25659.06
(R)-(-)-4-Penten-2-ol 95%
(R)-(-)-4-Penten-2-ol
Purity: 95%
CAS No.: 64584-92-5
Molecular Formula: C5H10O
Formula Weight: 86.13
Flash Point: 30°(86°F)
Refractive Index: 1.4240
Boiling Point: 115-116°
Optical Rotation: -5.2° (c=1 in chloroform)
UN No.: 1987
MDL No.: MFCD03701536
Formel:
C₅H₁₀O Siedepunkt: 115…116 °C Dichte: 0,837 Flammpunkt: 30 °C (86 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD03701536 CAS-Nummer: 64584-92-5 UN: 1987 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...