(3,5-Dimethoxyphenyl)essigsäure ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 2-(3,5-Dimethoxyphenyl)acetic acid

B25672.03 B25672.06
B25672.03EA 49 EUR
B25672.03 B25672.06
(3,5-Dimethoxyphenyl)essigsäure ≥98%
(3,5-Dimethoxyphenyl)essigsäure

Purity: 98% CAS No.: 4670-10-4 Molecular Formula: C10H12O4 Formula Weight: 196.20 Melting Point: 102-104° MDL No.: MFCD00016827

Formel: (C₆H₃(OCH₃)₂)CH₂COOH
Molecular Weight: 196.2 g/mol
Schmelzpunkt: 102…104 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00016827
CAS-Nummer: 4670-10-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR