2,6-Diacetylpyridin ≥99%

Lieferant: Thermo Fisher Scientific
Achtung

B25678.03 B25678.06 B25678.14
B25678.03EA 47.6 EUR
B25678.03 B25678.06 B25678.14
2,6-Diacetylpyridin ≥99%
2,6-Diacetylpyridin

Purity: 99% CAS No.: 1129-30-2 Molecular Formula: C9H9NO2 Formula Weight: 163.18 Melting Point: 78-82° MDL No.: MFCD00006304

Formel: C₉H₉NO₂
Molecular Weight: 163.18 g/mol
Schmelzpunkt: 78…82 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00006304
CAS-Nummer: 1129-30-2
EINECS: 214-442-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR