2-Brom-5-nitrobenzoesäure ≥98%

Lieferant: Thermo Scientific
Achtung

B25704.06
B25704.06EA 63.4 EUR
B25704.06
2-Brom-5-nitrobenzoesäure ≥98%
2-Bromo-5-nitrobenzoic acid

Purity: 98% CAS No.: 943-14-6 Molecular Formula: C7H4BrNO4 Formula Weight: 246.02 Melting Point: 178-181° Beilstein Reference: 980242

Formel: C₇H₄BrNO₄
Molecular Weight: 246,02 g/mol
Siedepunkt: 371 °C (1013 hPa)
Schmelzpunkt: 178…181 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00134558
CAS-Nummer: 943-14-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR