4-(Methylthio)benzoesäure ≥97%

Lieferant: Thermo Fisher Scientific
Achtung

B25706.03 B25706.06 B25706.14
B25706.03EA 40.8 EUR
B25706.03 B25706.06 B25706.14
4-(Methylthio)benzoesäure ≥97%
4-(Methylthio)benzoesäure

Purity: 97% CAS No.: 13205-48-6 Molecular Formula: C8H8O2S Formula Weight: 168.21 Melting Point: 193-196° Beilstein Reference: 2082809

Formel: C₈H₈O₂S
Schmelzpunkt: 193…196 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00002555
CAS-Nummer: 13205-48-6
EINECS: 236-171-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR