5-Fluorsalicylaldehyd ≥98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
5-Fluoro-2-hydroxybenzaldehyde
B25724.03
B25724.03EA
56
EUR
B25724.03
5-Fluorsalicylaldehyd ≥98%
5-Fluorsalicylaldehyd
Purity: 98+%
CAS No.: 347-54-6
Molecular Formula: C7H5FO2
Formula Weight: 140.11
Melting Point: 82-85°
MDL No.: MFCD01090997
Formel:
C₇H₅FO₂ Molecular Weight: 140,11 g/mol Schmelzpunkt: 82…85 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01090997 CAS-Nummer: 347-54-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...