2-Anilino-4,6-di(4-morpholinyl)-1,3,5-triazin ≥97%

Lieferant: Thermo Scientific
Achtung

B25741.03
B25741.03EA 161 EUR
B25741.03
2-Anilino-4,6-di(4-morpholinyl)-1,3,5-triazin ≥97%
2-Anilino-4,6-di(4-morpholinyl)-1,3,5-triazin

Purity: 97% CAS No.: 93438-27-8 Molecular Formula: C17H22N6O2 Formula Weight: 342.40 Melting Point: 171-174° MDL No.: MFCD00272422

Formel: C₁₇H₂₂N₆O₂
Molecular Weight: 342,40 g/mol
Schmelzpunkt: 171…174 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00272422
CAS-Nummer: 93438-27-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR